siRNA Details
Gene Symbol: Olfr181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC125397.1 | - | 1 | 802 | 40129912 | |
BC131916.1 | - | 1 | 802 | 40130193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
