siRNA Details
Gene Symbol: Olfr1410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146491.1 | NP_666702.1 | 1 | 315 | ||
GenBank mRNA | BC106991.1 | - | 1 | 315 | 40044064 | |
BC106992.1 | - | 1 | 315 | 40044072 |
Gene Symbol: Olfr1413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147037.2 | NP_667248.1 | 1 | 363 | ||
GenBank mRNA | BC116898.1 | - | 1 | 363 | 40126217 |

More Information
Other Species Targeted

Gene Ontology Categories:
