siRNA Details
Gene Symbol: Olfr679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147044.1 | NP_667255.1 | 1 | 260 |
Gene Symbol: Olfr681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207557.2 | NP_997440.2 | 1 | 260 | ||
GenBank mRNA | BC104298.1 | - | 1 | 260 | 40043142 | |
BC104299.1 | - | 1 | 260 | 40043148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
