siRNA Details
Gene Symbol: TUBB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001069.2 | NP_001060.1 | 2 | 160 | ||
NM_001310315.1 | NP_001297244.1 | 2 | 160 | |||
GenBank mRNA | AK312621.1 | - | 2 | 155 | ||
AY159127.1 | - | 2 | 74 | |||
BC001194.1 | - | 2 | 123 | 3355410 | ||
BC018780.1 | - | 2 | 134 | 4870519 | ||
BC127873.1 | - | 1 | 113 | |||
BC141924.1 | - | 1 | 113 | |||
BC141925.1 | - | 1 | 113 | |||
BM846690.1 | - | 2 | 154 | |||
BP381259.1 | - | 2 | 184 | |||
CR541958.1 | - | 2 | 74 | |||
X79535.1 | - | 2 | 137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
