siRNA Details
Gene Symbol: Olfr1062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147078.2 | NP_667289.2 | 1 | 705 |
Gene Symbol: Olfr1057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207563.2 | NP_997446.1 | 1 | 705 |

More Information
Other Species Targeted

Gene Ontology Categories:
