siRNA Details
Gene Symbol: Ddx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040187.1 | NP_001035277.1 | 13 | 2076 | ||
NM_152806.3 | NP_690019.2 | 2 | 1378 | |||
NM_199080.2 | NP_951062.1 | 13 | 2082 | |||
GenBank mRNA | AK027954.1 | - | 2 | 228 | ||
AK151061.1 | - | 13 | 1857 | |||
AK152749.1 | - | 13 | 1858 | |||
AK152831.1 | - | 13 | 1857 | |||
AK152910.1 | - | 2 | 317 | |||
BC027758.1 | - | 2 | 615 | |||
BC031802.1 | - | 1 | 273 | |||
BC038378.1 | - | 2 | 964 | |||
BC096036.1 | - | 13 | 2076 | 30549388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
