siRNA Details
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276340.1 | NP_001263269.1 | 2 | 412 | ||
GenBank mRNA | AB079133.1 | - | 2 | 246 | ||
BB612294.1 | - | 2 | 222 | |||
BG086617.2 | - | 2 | 227 |

More Information
Other Species Targeted

Gene Ontology Categories:
