siRNA Details
Gene Symbol: Egfl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152922.3 | NP_690886.3 | 2 | 165 | ||
GenBank mRNA | AK148723.1 | - | 2 | 166 | ||
BC141312.1 | - | 2 | 134 | 9055924 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
