siRNA Details
Gene Symbol: Slc37a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242427.1 | NP_001229356.1 | 20 | 2449 | ||
NM_153062.2 | NP_694702.1 | 20 | 2029 | |||
GenBank mRNA | AK033656.1 | - | 20 | 1918 | ||
AK143636.1 | - | 20 | 2455 | |||
AK153706.1 | - | 20 | 2452 | |||
AK163990.1 | - | 19 | 1632 | |||
BC027294.1 | - | 20 | 2029 | 3985469 | ||
BC029208.1 | - | 12 | 849 |
