siRNA Details
Gene Symbol: Slc9a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB089793.1 | - | 5 | 444 | ||
AF482993.1 | - | 4 | 408 | |||
AK037377.1 | - | 5 | 431 | |||
AK049410.1 | - | 5 | 440 | |||
AK051791.1 | - | 4 | 359 | |||
AK143281.1 | - | 5 | 413 | |||
AK173069.1 | - | 5 | 780 | |||
BC030879.1 | - | 5 | 443 | 4241988 | ||
BC058947.1 | - | 5 | 414 | 5702781 |
