siRNA Details
Gene Symbol: Ddx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040187.1 | NP_001035277.1 | 13 | 2159 | ||
NM_152806.3 | NP_690019.2 | 2 | 1461 | |||
NM_199080.2 | NP_951062.1 | 13 | 2165 | |||
GenBank mRNA | AK027954.1 | - | 2 | 311 | ||
AK151061.1 | - | 13 | 1940 | |||
AK152749.1 | - | 13 | 1941 | |||
AK152831.1 | - | 13 | 1940 | |||
AK152910.1 | - | 2 | 400 | |||
BC027758.1 | - | 2 | 698 | |||
BC031802.1 | - | 1 | 356 | |||
BC038378.1 | - | 2 | 1047 | |||
BC096036.1 | - | 13 | 2159 | 30549388 |

More Information
Other Species Targeted

Gene Ontology Categories:
