siRNA Details
Gene Symbol: Kdm5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152895.2 | NP_690855.2 | 3 | 512 | ||
GenBank mRNA | AK008738.1 | - | 3 | 465 | ||
AK041304.1 | - | 3 | 519 | |||
AK163481.1 | - | 3 | 484 | |||
AK220451.1 | - | 3 | 498 | |||
AY082429.1 | - | 3 | 495 | |||
AY082430.1 | - | 3 | 495 | |||
BC048180.1 | - | 3 | 495 | 6826125 | ||
BC057318.1 | - | 3 | 495 | 6826125 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK086430.1 | - | 14 | Chr. - - [-] on Build |
