siRNA Details
Gene Symbol: Ddx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040187.1 | NP_001035277.1 | 13 | 2731 | ||
NM_152806.3 | NP_690019.2 | 2 | 2033 | |||
NM_199080.2 | NP_951062.1 | 13 | 2737 | |||
GenBank mRNA | AK151061.1 | - | 13 | 2512 | ||
AK152749.1 | - | 13 | 2515 | |||
AK152831.1 | - | 13 | 2512 | |||
AK152910.1 | - | 2 | 972 | |||
BC027758.1 | - | 2 | 1270 | |||
BC031802.1 | - | 1 | 928 | |||
BC038378.1 | - | 2 | 1619 | |||
BC096036.1 | - | 13 | 2731 | 30549388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
