siRNA Details
Gene Symbol: Tmem191c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177473.3 | NP_803424.2 | 9 | 691 | ||
GenBank mRNA | AB041601.1 | - | 6 | 1008 | ||
BC058760.1 | - | 5 | 786 | |||
BC098238.1 | - | 5 | 318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
