siRNA Details
Gene Symbol: E130308A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015681.2 | NP_001015681.1 | 2 | 428 | ||
NM_153158.5 | NP_694798.1 | 2 | 428 | |||
GenBank mRNA | AK047478.1 | - | 2 | 429 | ||
AK047601.1 | - | 2 | 397 | |||
AK081604.1 | - | 2 | 312 | |||
BC017649.1 | - | 2 | 195 | 4924105 | ||
BC037070.1 | - | 2 | 246 | 5004205 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006238214, XM_006238215, XM_008763745.
Gene Ontology Categories:
