siRNA Details
Gene Symbol: Clec4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204241.1 | NP_001191170.1 | 5 | 423 | ||
NM_153197.4 | NP_694877.2 | 4 | 559 | |||
GenBank mRNA | AK014135.1 | - | 4 | 459 | ||
AK156040.1 | - | 4 | 559 | |||
AY397674.1 | - | 4 | 370 | |||
BC034893.1 | - | 5 | 423 | 1496162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
