siRNA Details
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080448.4 | NP_536696.4 | 22 | 7618 | ||
GenBank mRNA | AF481964.1 | - | 1 | 503 | ||
AK122276.1 | - | 18 | 5899 | |||
AK163776.1 | - | 1 | 968 | |||
BC055702.1 | - | 1 | 1153 | |||
BC069855.1 | - | 1 | 281 | |||
BC128297.1 | - | 1 | 232 | |||
BC128298.1 | - | 1 | 232 | |||
BC128300.1 | - | 1 | 232 | |||
BC152363.1 | - | 1 | 237 | |||
BC152364.1 | - | 1 | 237 | |||
BC152366.1 | - | 1 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
