siRNA Details
Gene Symbol: St7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253702.1 | NP_001240631.1 | 2 | 862 | ||
NM_001253703.1 | NP_001240632.1 | 2 | 862 | |||
NM_153091.3 | NP_694731.1 | 2 | 862 | |||
GenBank mRNA | AB084518.1 | - | 2 | 276 | ||
AK033865.1 | - | 2 | 294 | |||
AK144075.1 | - | 2 | 302 | |||
AK148099.1 | - | 2 | 564 | |||
AK172102.1 | - | 2 | 863 | |||
AK172218.1 | - | 2 | 863 | |||
BC024473.1 | - | 2 | 253 | |||
BC058781.1 | - | 2 | 287 | 6311899 | ||
BC107394.2 | - | 2 | 287 | 40058131 | ||
BC145950.1 | - | 2 | 354 | 40131253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
