siRNA Details
Gene Symbol: PLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190801.1 | NP_001177730.1 | 3 | 259 | ||
NM_014264.4 | NP_055079.3 | 3 | 436 | |||
XM_005262701.2 | XP_005262758.1 | 3 | 398 | |||
XM_017007662.1 | XP_016863151.1 | 3 | 595 | |||
XM_017007663.1 | XP_016863152.1 | 3 | 596 | |||
GenBank mRNA | AB006972.1 | - | 3 | 350 | ||
AB741646.1 | - | 3 | 162 | |||
AK302858.1 | - | 3 | 259 | |||
AK314238.1 | - | 3 | 404 | |||
BC036023.1 | - | 3 | 404 | 5273226 | ||
BP328161.1 | - | 3 | 408 | |||
DC402409.1 | - | 3 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
