siRNA Details
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162868.1 | NP_001156340.1 | 6, 7 | 2170 | ||
NM_001162869.1 | NP_001156341.1 | 7, 8 | 2305 | |||
NM_153140.2 | NP_694780.1 | 4, 5 | 561 | |||
GenBank mRNA | AB093257.1 | - | 7, 8 | 1746 | ||
AK052775.1 | - | 5, 6 | 654 | |||
AK083658.1 | - | 6, 7 | 1854 | |||
AK085516.1 | - | 4, 5 | 562 | |||
AK128985.1 | - | 4, 5 | 561 | |||
BC023364.1 | - | 4, 5 | 552 | |||
BC037132.1 | - | 4, 5 | 601 | |||
CF170100.1 | - | 4, 5 | 460 |

More Information
Other Species Targeted

Gene Ontology Categories:
