siRNA Details
Gene Symbol: Kctd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153143.4 | NP_694783.1 | 1 | 963 | ||
GenBank mRNA | AF465352.1 | - | 1 | 930 | ||
AK039558.1 | - | 1385 | ||||
AK161602.1 | - | 1 | 958 | |||
AK170258.1 | - | 1 | 857 | |||
BC052502.1 | - | 1 | 211 | 5119264 | ||
BC052734.1 | - | 1 | 914 | 6816736 | ||
BC063052.1 | - | 1 | 914 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108831.
Gene Ontology Categories:
