siRNA Details
Gene Symbol: Pkhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153179.3 | NP_694819.2 | 6 | 630 | ||
GenBank mRNA | AK052698.1 | - | 6 | 639 | ||
AY130764.1 | - | 6 | 622 | |||
AY438374.1 | - | 6 | 504 |
