siRNA Details
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080448.4 | NP_536696.4 | 22 | 7638 | ||
GenBank mRNA | AF481964.1 | - | 1 | 523 | ||
AK122276.1 | - | 18 | 5919 | |||
AK163776.1 | - | 1 | 988 | |||
BC055702.1 | - | 1 | 1173 | |||
BC069855.1 | - | 1 | 301 | |||
BC128297.1 | - | 1 | 252 | |||
BC128298.1 | - | 1 | 252 | |||
BC128300.1 | - | 1 | 252 | |||
BC152363.1 | - | 1 | 257 | |||
BC152364.1 | - | 1 | 257 | |||
BC152366.1 | - | 1 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
