siRNA Details
Gene Symbol: Atf7ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153123.2 | NP_694763.2 | 11 | 2259 | ||
GenBank mRNA | AK145634.1 | - | 11 | 2217 | ||
BC018510.1 | - | 1 | 707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
