siRNA Details
Gene Symbol: Hecw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081348.3 | NP_001074817.3 | 18, 19 | 3873 | ||
GenBank mRNA | AB083710.1 | - | 16, 17 | 3376 | ||
AK029861.1 | - | 10, 11 | 2477 | |||
AK172932.1 | - | 9, 10 | 2158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
