siRNA Details
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162868.1 | NP_001156340.1 | 14 | 4489 | ||
NM_001162869.1 | NP_001156341.1 | 15 | 4624 | |||
NM_153140.2 | NP_694780.1 | 12 | 2880 | |||
GenBank mRNA | AB093257.1 | - | 15 | 4065 | ||
AK083658.1 | - | 14 | 4178 | |||
AK085516.1 | - | 11 | 2920 | |||
AK128985.1 | - | 12 | 2880 | |||
BC023364.1 | - | 12 | 2871 | |||
BC037132.1 | - | 12 | 2920 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_002727755, XM_006246071, XM_220262, BC081997.
Gene Ontology Categories:
