siRNA Details
Gene Symbol: Cnot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205226.1 | NP_001192155.1 | 35, 36 | 5282 | ||
NM_153164.3 | NP_694804.3 | 35, 36 | 5288 | |||
NM_178078.2 | NP_835179.1 | 35, 36 | 5288 | |||
GenBank mRNA | AK129258.1 | - | 2248 | |||
BC018281.1 | - | 8, 9 | 1259 | |||
BC058249.1 | - | 35, 36 | 5186 | |||
BC157948.1 | - | 35, 36 | 5201 | 9088123 | ||
BC158073.1 | - | 35, 36 | 5207 | 9007370 | ||
BC172105.1 | - | 35, 36 | 5207 | 9054391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
