siRNA Details
Gene Symbol: Ago2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153178.4 | NP_694818.3 | 19 | 3068 | ||
GenBank mRNA | AK220193.1 | - | 16 | 2433 | ||
BC023279.1 | - | 11 | 1769 | |||
BC024857.1 | - | 12 | 1913 | |||
BC056639.1 | - | 9 | 1635 | |||
BC064741.1 | - | 14 | 2185 | |||
BC096465.1 | - | 19 | 2973 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
