siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153399.2 | NP_700448.2 | 5, 6 | 745 | ||
GenBank mRNA | AF535143.1 | - | 5, 6 | 745 | ||
AK044459.1 | - | 5, 6 | 732 |

More Information
Other Species Targeted

Gene Ontology Categories:
