siRNA Details
Gene Symbol: Rtn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153457.7 | NP_703187.2 | 2 | 865 | ||
GenBank mRNA | AB074899.1 | - | 2 | 362 | ||
AK132364.1 | - | 2 | 865 | |||
BC030455.1 | - | 2 | 417 | 5401661 | ||
BC053926.1 | - | 2 | 493 | 6312477 | ||
BC058579.1 | - | 2 | 527 | 6490092 | ||
CT010392.1 | - | 2 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
