siRNA Details
Gene Symbol: Marf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081154.2 | NP_001074623.1 | 9 | 2269 | ||
GenBank mRNA | AK035704.1 | - | 1923 | |||
AK220341.1 | - | 6 | 3933 | |||
BC031575.1 | - | 2 | 441 | |||
BC069852.1 | - | 5 | 955 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
