siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153399.2 | NP_700448.2 | 28 | 3970 | ||
GenBank mRNA | AF535143.1 | - | 28 | 3970 | ||
AK032502.1 | - | 15 | 2167 | |||
AK122550.3 | - | 15 | 2400 | |||
AK148504.1 | - | 14 | 2070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
