siRNA Details
Gene Symbol: Pomt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153415.3 | NP_700464.2 | 13 | 1896 | ||
GenBank mRNA | AF246235.1 | - | 13 | 1500 | ||
AK043573.1 | - | 13 | 1597 | |||
AK043713.1 | - | 13 | 1438 | |||
AK050915.1 | - | 13 | 1465 | |||
AK076813.1 | - | 13 | 1898 | |||
AK214879.1 | - | 1 | 17 | |||
BC052045.1 | - | 13 | 1468 | 6402328 | ||
BC062965.1 | - | 13 | 1468 | |||
BC079589.1 | - | 13 | 1466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
