siRNA Details
Gene Symbol: Rrnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC012224.1 | - | 2, 3 | 397 | ||
BC023814.1 | - | 1, 2 | 345 | 5353932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
