siRNA Details
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193424.1 | NP_001180353.1 | 6 | 3047 | ||
NM_001193425.1 | NP_001180354.1 | 6 | 3005 | |||
NM_001193426.1 | NP_001180355.1 | 6 | 2507 | |||
NM_001193427.1 | NP_001180356.1 | 6 | 2465 | |||
NM_024670.3 | NP_078946.1 | 5 | 2992 | |||
NR_034181.1 | 5 | 2901 | ||||
XM_011519662.2 | XP_011517964.1 | 6 | 3036 | |||
GenBank mRNA | AK027067.1 | - | 5 | 2798 | ||
BC007754.2 | - | 5 | 2992 | 4109807 |
Gene Symbol: NINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093569.1 | - | 6 | 2544 |
Gene Symbol: NME6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311152.1 | - | 2 | 677 |
Gene Symbol: PDE4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000923.4 | NP_000914.2 | 16 | 3511 | ||
NM_001098818.2 | NP_001092288.1 | 15 | 3236 | |||
NM_001098819.2 | NP_001092289.1 | 15 | 3289 | |||
NR_040546.1 | 14 | 3186 | ||||
XM_017026866.1 | XP_016882355.1 | 16 | 3520 | |||
GenBank mRNA | Z46632.1 | - | 15 | 3383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
