siRNA Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153787.2 | NP_722482.1 | 3, 4 | 362 | ||
GenBank mRNA | AK088741.1 | - | 3, 4 | 303 | ||
AK141834.1 | - | 3, 4 | 315 | |||
BC034300.1 | - | 3, 4 | 294 | 4507876 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006227654, XM_006227656.
Gene Ontology Categories:
