siRNA Details
Gene Symbol: Arid4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081195.1 | NP_001074664.1 | 3 | 333 | ||
GenBank mRNA | AK042186.1 | - | 2 | 522 | ||
AK050594.1 | - | 3 | 273 | |||
AK171991.1 | - | 1 | 2343 | |||
AY116674.1 | - | 3 | 220 | |||
BC025436.1 | - | 3 | 321 | |||
BC058209.1 | - | 3 | 250 | |||
BC080780.1 | - | 3 | 268 | |||
BC157975.1 | - | 2 | 135 | 9007297 | ||
BC158109.1 | - | 2 | 135 | 9007390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
