siRNA Details
Gene Symbol: Zfp628 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170759.2 | NP_739565.2 | 3 | 768 | ||
GenBank mRNA | AF435832.1 | - | 1 | 472 | ||
AK048291.1 | - | 3 | 735 | |||
AK155214.1 | - | 3 | 768 | |||
BC056945.1 | - | 3 | 701 | |||
BC062973.1 | - | 3 | 701 |
