siRNA Details
Gene Symbol: Ttc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003910.2 | NP_001003910.2 | 2 | 805 | ||
NM_001003939.2 | NP_001003939.2 | 3 | 914 | |||
NM_153595.3 | NP_705823.3 | 1 | 598 | |||
GenBank mRNA | AK133195.1 | - | 2 | 745 | ||
BC030307.1 | - | 3 | 892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
