siRNA Details
Gene Symbol: Rnf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153762.3 | NP_717095.2 | 1 | 2084 | ||
GenBank mRNA | AK033164.1 | - | 1 | 2538 | ||
AK157821.1 | - | 1 | 2533 | |||
BC004739.1 | - | 1 | 2130 | 3501511 | ||
BC063251.1 | - | 1 | 2084 | 6413980 | ||
BC086757.1 | - | 2 | 1624 | |||
BC111036.1 | - | 2 | 2068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
