siRNA Details
Gene Symbol: Cdkl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166655.1 | NP_001160127.1 | 9 | 1703 | ||
NM_001166657.1 | NP_001160129.1 | 8 | 1540 | |||
GenBank mRNA | AK050990.1 | - | 9 | 1700 | ||
AK090045.1 | - | 9 | 1487 | |||
BC028871.1 | - | 8 | 1412 | 4241862 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
