siRNA Details
Gene Symbol: Plekhg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080928.1 | - | 12 | 3418 | ||
BC030417.1 | - | 10 | 3110 | |||
BC049264.1 | - | 10 | 3099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
