siRNA Details
Gene Symbol: Arid4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081195.1 | NP_001074664.1 | 11 | 1128 | ||
GenBank mRNA | AK050594.1 | - | 10 | 935 | ||
AY116674.1 | - | 11 | 1014 | |||
BC025436.1 | - | 11 | 1116 | |||
BC058209.1 | - | 11 | 1045 | |||
BC080780.1 | - | 11 | 1063 | |||
BC157975.1 | - | 10 | 930 | 9007297 | ||
BC158109.1 | - | 10 | 930 | 9007390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
