siRNA Details
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172146.2 | NP_742158.1 | 2, 3 | 697 | ||
GenBank mRNA | AK147750.1 | - | 2, 3 | 347 | ||
AK148034.1 | - | 2, 3 | 301 | |||
AK159141.1 | - | 3, 4 | 847 | |||
AK159859.1 | - | 3, 4 | 847 | |||
AK160542.1 | - | 2, 3 | 322 | |||
AK164506.1 | - | 2, 3 | 311 | |||
AK167057.1 | - | 2, 3 | 336 | |||
BC006053.1 | - | 2, 3 | 284 | |||
BC023841.1 | - | 2, 3 | 324 | 5346105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
