siRNA Details
Gene Symbol: Plb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081407.1 | NP_001074876.1 | 59 | 4504 | ||
NM_172147.2 | NP_742159.1 | 5 | 414 | |||
GenBank mRNA | AK087596.1 | - | 415 | |||
BC033606.1 | - | 5 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
