siRNA Details
Gene Symbol: Zdhhc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172151.4 | NP_742163.4 | 11 | 2424 | ||
GenBank mRNA | AK032069.1 | - | 2 | 414 | ||
AK213447.1 | - | 1 | 29 | |||
AY894892.1 | - | 11 | 2424 | |||
BC004683.1 | - | 1 | 235 | |||
BC055694.1 | - | 2 | 721 | |||
BC083154.1 | - | 1 | 345 | |||
BC106166.1 | - | 1 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
