siRNA Details
Gene Symbol: Nup214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172268.2 | NP_758472.2 | 15, 16 | 2228 | ||
GenBank mRNA | AK122189.1 | - | 15, 16 | 2197 | ||
AK162214.1 | - | 924 | ||||
BC039282.1 | - | 5, 6 | 585 | 4976266 |

More Information
Other Species Targeted

Gene Ontology Categories:
