siRNA Details
Gene Symbol: Zmynd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252584.2 | NP_001239513.1 | 3, 4 | 453 | ||
NM_001252587.2 | NP_001239516.1 | 4, 5 | 465 | |||
NM_001281926.1 | NP_001268855.1 | 3, 4 | 352 | |||
GenBank mRNA | AK147620.1 | - | 3, 4 | 353 | ||
AK155864.1 | - | 3, 4 | 258 | |||
BC023300.1 | - | 3, 4 | 320 | |||
BC063267.1 | - | 3, 4 | 246 | |||
CN536818.1 | - | 3, 4 | 488 | |||
CN670238.1 | - | 4, 5 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
