siRNA Details
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162977.1 | NP_001156449.1 | 30 | 4094 | ||
GenBank mRNA | AK134303.1 | - | 17 | 2088 | ||
AK173032.1 | - | 13 | 1573 | |||
AK186305.1 | - | 3 | 197 | |||
BC031402.1 | - | 10 | 1132 | |||
BC039980.1 | - | 10 | 1143 | |||
BC058571.1 | - | 9 | 1036 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
