siRNA Details
Gene Symbol: 2210018M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172280.2 | NP_758484.2 | 3 | 221 | ||
GenBank mRNA | AK032985.1 | - | 3 | 248 | ||
AK137691.1 | - | 3 | 437 | |||
BC039956.1 | - | 3 | 212 | |||
BC066050.1 | - | 3 | 198 | |||
BC089304.1 | - | 3 | 221 | 6854147 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006229812, XM_006229802, XM_006229809, XM_006229810, XM_006229813, and others.
Gene Ontology Categories:
